Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCACCAACTCTTTTGAGCCTAA[A/G]CTCTAACTAATCAGAGTTGACAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611380 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIP2C PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DIP2C - disco interacting protein 2 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014974.2 | 5363 | UTR 3 | NP_055789.1 | |||
XM_005252426.3 | 5363 | UTR 3 | XP_005252483.1 | |||
XM_005252427.3 | 5363 | UTR 3 | XP_005252484.1 | |||
XM_005252428.4 | 5363 | UTR 3 | XP_005252485.1 | |||
XM_005252430.2 | 5363 | UTR 3 | XP_005252487.2 | |||
XM_011519428.2 | 5363 | UTR 3 | XP_011517730.1 | |||
XM_011519429.2 | 5363 | UTR 3 | XP_011517731.1 | |||
XM_011519430.2 | 5363 | UTR 3 | XP_011517732.1 | |||
XM_011519431.2 | 5363 | UTR 3 | XP_011517733.1 | |||
XM_011519432.1 | 5363 | UTR 3 | XP_011517734.1 | |||
XM_011519433.2 | 5363 | UTR 3 | XP_011517735.1 | |||
XM_017015977.1 | 5363 | UTR 3 | XP_016871466.1 |