Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAAGCAAGTCTTGGACAGTAAG[G/T]GCGCCTTCTTCGGAGCCCTCTGCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601662 | |||||||||||||||||||||||||||||||||||
Literature Links: |
ALCAM PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
ALCAM - activated leukocyte cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243280.1 | Intron | NP_001230209.1 | ||||
NM_001243281.1 | Intron | NP_001230210.1 | ||||
NM_001243283.1 | Intron | NP_001230212.1 | ||||
NM_001627.3 | Intron | NP_001618.2 |
Set Membership: |
Validated |