Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAAACACGAGATAAGCTTTGGC[A/G]AGAATGTAGAGAAAGGAAACTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611700 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SVOPL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SVOPL - SVOP like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139456.1 | Intron | NP_001132928.1 | ||||
NM_174959.3 | Intron | NP_777619.1 | ||||
XM_005250143.3 | Intron | XP_005250200.1 | ||||
XM_011515797.2 | Intron | XP_011514099.1 | ||||
XM_017011746.1 | Intron | XP_016867235.1 | ||||
XM_017011747.1 | Intron | XP_016867236.1 | ||||
XM_017011748.1 | Intron | XP_016867237.1 |
Set Membership: |
HapMap |