Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATTCTGGTTCACAGAATTTCTA[G/T]GGCAGCTTTCAGATGAAACCATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605569 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR83 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPR83 - G protein-coupled receptor 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016540.3 | 3523 | UTR 3 | NP_057624.3 | |||
XM_011542559.2 | 3523 | UTR 3 | XP_011540861.1 |
Set Membership: |
HapMap |