Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCCTGCACCCTCACACCATAGC[C/G]CTGGCACCTGCCCCTCTTGCCCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986255 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107986255 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK32B - serine/threonine kinase 32B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306082.1 | Intron | NP_001293011.1 | ||||
NM_018401.2 | Intron | NP_060871.1 | ||||
XM_005247982.3 | Intron | XP_005248039.1 | ||||
XM_005247983.4 | Intron | XP_005248040.1 | ||||
XM_011513497.1 | Intron | XP_011511799.1 | ||||
XM_011513501.2 | Intron | XP_011511803.1 | ||||
XM_011513504.1 | Intron | XP_011511806.1 | ||||
XM_017008374.1 | Intron | XP_016863863.1 | ||||
XM_017008375.1 | Intron | XP_016863864.1 | ||||
XM_017008376.1 | Intron | XP_016863865.1 | ||||
XM_017008377.1 | Intron | XP_016863866.1 |
Set Membership: |
HapMap |