Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGGCCCCACACACATAGCATCC[A/G]ACACAACAGCAGGCACAAAGGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603336 MIM: 614912 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAH6 - dynein axonemal heavy chain 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRABD2A - TraB domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080824.2 | Intron | NP_001074293.1 | ||||
NM_001277053.1 | Intron | NP_001263982.1 | ||||
NM_001307978.1 | Intron | NP_001294907.1 | ||||
XM_011532504.1 | Intron | XP_011530806.1 |
Set Membership: |
HapMap |