Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCCATGGGGTTCCCAAGGTGG[C/T]GTGGAGGCCCATGCAGCTCCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611746 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373051 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105373051 - uncharacterized LOC105373051 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCUBE1 - signal peptide, CUB domain and EGF like domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173050.3 | 3180 | UTR 3 | NP_766638.2 | |||
XM_005261752.1 | 3180 | UTR 3 | XP_005261809.1 | |||
XM_011530389.2 | 3180 | UTR 3 | XP_011528691.1 | |||
XM_011530390.2 | 3180 | UTR 3 | XP_011528692.1 |
Set Membership: |
HapMap |