Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGACTCCCCAGGCCTCCTGGAAG[G/A]GTGTGGCTGCTGCTGCTCCCCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612395 MIM: 601987 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHKB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHKB - choline kinase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005198.4 | Intron | NP_005189.2 |
CHKB-AS1 - CHKB antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHKB-CPT1B - CHKB-CPT1B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPT1B - carnitine palmitoyltransferase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145134.1 | Intron | NP_001138606.1 | ||||
NM_001145135.1 | Intron | NP_001138607.1 | ||||
NM_001145137.1 | Intron | NP_001138609.1 | ||||
NM_004377.3 | Intron | NP_004368.1 | ||||
NM_152245.2 | Intron | NP_689451.1 | ||||
NM_152246.2 | Intron | NP_689452.1 |
Set Membership: |
Validated |