Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607826 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCYL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AHCYL1 - adenosylhomocysteinase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRIP1 - striatin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270768.1 | Intron | NP_001257697.1 | ||||
NM_033088.3 | Intron | NP_149079.2 | ||||
XM_006710995.2 | Intron | XP_006711058.1 | ||||
XM_017002629.1 | Intron | XP_016858118.1 | ||||
XM_017002630.1 | Intron | XP_016858119.1 |