Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600723 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MPP2 - membrane palmitoylated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278370.1 | 3084 | UTR 3 | NP_001265299.1 | |||
NM_001278371.1 | 3084 | UTR 3 | NP_001265300.1 | |||
NM_001278372.1 | 3084 | UTR 3 | NP_001265301.1 | |||
NM_001278373.1 | 3084 | UTR 3 | NP_001265302.1 | |||
NM_001278374.1 | 3084 | UTR 3 | NP_001265303.1 | |||
NM_001278375.1 | 3084 | UTR 3 | NP_001265304.1 | |||
NM_001278376.2 | 3084 | UTR 3 | NP_001265305.2 | |||
NM_001278381.1 | 3084 | UTR 3 | NP_001265310.1 | |||
NM_005374.4 | 3084 | UTR 3 | NP_005365.4 | |||
XM_011524827.2 | 3084 | UTR 3 | XP_011523129.2 |
Set Membership: |
HapMap |