Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605265 MIM: 118496 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AVEN PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AVEN - apoptosis and caspase activation inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020371.2 | Intron | NP_065104.1 | ||||
XM_011521818.2 | Intron | XP_011520120.1 | ||||
XM_011521819.2 | Intron | XP_011520121.1 | ||||
XM_011521820.1 | Intron | XP_011520122.1 |
CHRM5 - cholinergic receptor muscarinic 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320917.1 | Intron | NP_001307846.1 | ||||
NM_012125.3 | Intron | NP_036257.1 |
Set Membership: |
HapMap |