Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614236 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC38A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC38A7 - solute carrier family 38 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308384.1 | 2354 | UTR 3 | NP_001295313.1 | |||
NM_018231.2 | 2354 | UTR 3 | NP_060701.1 | |||
XM_017023393.1 | 2354 | UTR 3 | XP_016878882.1 | |||
XM_017023394.1 | 2354 | UTR 3 | XP_016878883.1 | |||
XM_017023395.1 | 2354 | UTR 3 | XP_016878884.1 | |||
XM_017023396.1 | 2354 | UTR 3 | XP_016878885.1 | |||
XM_017023397.1 | 2354 | Intron | XP_016878886.1 | |||
XM_017023398.1 | 2354 | Intron | XP_016878887.1 | |||
XM_017023399.1 | 2354 | Intron | XP_016878888.1 |
Set Membership: |
HapMap |