Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616908 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTCHD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PTCHD4 - patched domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013732.3 | 23206 | Intron | NP_001013754.3 | |||
NM_207499.2 | 23206 | Intron | NP_997382.2 | |||
XM_011514639.2 | 23206 | UTR 3 | XP_011512941.1 | |||
XM_017010891.1 | 23206 | UTR 3 | XP_016866380.1 | |||
XM_017010892.1 | 23206 | UTR 3 | XP_016866381.1 | |||
XM_017010893.1 | 23206 | UTR 3 | XP_016866382.1 | |||
XM_017010894.1 | 23206 | UTR 3 | XP_016866383.1 | |||
XM_017010895.1 | 23206 | UTR 3 | XP_016866384.1 | |||
XM_017010896.1 | 23206 | Intron | XP_016866385.1 |
Set Membership: |
HapMap Validated |