Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605938 MIM: 602259 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIGP - phosphatidylinositol glycan anchor biosynthesis class P | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC3 - tetratricopeptide repeat domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320703.1 | Intron | NP_001307632.1 | ||||
NM_001320704.1 | Intron | NP_001307633.1 | ||||
NM_003316.3 | Intron | NP_003307.3 | ||||
XM_005261048.1 | Intron | XP_005261105.1 | ||||
XM_005261049.1 | Intron | XP_005261106.1 | ||||
XM_005261050.4 | Intron | XP_005261107.1 | ||||
XM_005261052.1 | Intron | XP_005261109.1 | ||||
XM_005261054.3 | Intron | XP_005261111.1 | ||||
XM_005261055.1 | Intron | XP_005261112.1 | ||||
XM_005261056.2 | Intron | XP_005261113.1 | ||||
XM_011529739.2 | Intron | XP_011528041.1 | ||||
XM_011529740.2 | Intron | XP_011528042.1 | ||||
XM_011529742.2 | Intron | XP_011528044.2 | ||||
XM_017028458.1 | Intron | XP_016883947.1 | ||||
XM_017028459.1 | Intron | XP_016883948.1 | ||||
XM_017028460.1 | Intron | XP_016883949.1 | ||||
XM_017028461.1 | Intron | XP_016883950.1 | ||||
XM_017028462.1 | Intron | XP_016883951.1 | ||||
XM_017028463.1 | Intron | XP_016883952.1 | ||||
XM_017028464.1 | Intron | XP_016883953.1 | ||||
XM_017028465.1 | Intron | XP_016883954.1 | ||||
XM_017028466.1 | Intron | XP_016883955.1 | ||||
XM_017028467.1 | Intron | XP_016883956.1 |
Set Membership: |
HapMap |