Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCGAAGCCGGAGGAGCCGGGC[A/G]TCTCCTCCCCGCCGGCCCGCGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBR3-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBR3-AS1 - CBR3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DOPEY2 - dopey family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320714.1 | Intron | NP_001307643.1 | ||||
NM_005128.3 | Intron | NP_005119.2 | ||||
XM_017028509.1 | Intron | XP_016883998.1 |