Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610633 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICU3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MICU3 - mitochondrial calcium uptake family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181723.2 | Intron | NP_859074.1 | ||||
XM_005273477.2 | Intron | XP_005273534.1 | ||||
XM_006716328.2 | Intron | XP_006716391.1 | ||||
XM_006716329.2 | Intron | XP_006716392.1 | ||||
XM_006716330.2 | Intron | XP_006716393.1 | ||||
XM_006716333.3 | Intron | XP_006716396.1 | ||||
XM_011544507.2 | Intron | XP_011542809.1 | ||||
XM_011544508.2 | Intron | XP_011542810.1 | ||||
XM_017013336.1 | Intron | XP_016868825.1 | ||||
XM_017013337.1 | Intron | XP_016868826.1 | ||||
XM_017013338.1 | Intron | XP_016868827.1 | ||||
XM_017013339.1 | Intron | XP_016868828.1 | ||||
XM_017013340.1 | Intron | XP_016868829.1 | ||||
XM_017013341.1 | Intron | XP_016868830.1 | ||||
XM_017013342.1 | Intron | XP_016868831.1 | ||||
XM_017013343.1 | Intron | XP_016868832.1 |
Set Membership: |
Validated |