Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGAACCAGCTGAGGCTGTGACC[A/C]CTCCAGCCACCTGCCCCAACCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609406 | ||||||||||||||||||||||||||||||||
Literature Links: |
PRR5 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PRR5 - proline rich 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017528.2 | Intron | NP_001017528.1 | ||||
NM_001017529.2 | Intron | NP_001017529.1 | ||||
NM_001017530.1 | Intron | NP_001017530.1 | ||||
NM_001198721.1 | Intron | NP_001185650.1 | ||||
NM_015366.3 | Intron | NP_056181.2 | ||||
NM_181333.3 | Intron | NP_851850.1 |
PRR5-ARHGAP8 - PRR5-ARHGAP8 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181334.5 | Intron | NP_851851.3 |
Set Membership: |
HapMap |