Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCGGAGGACATGGTGCGCCGTGT[A/C]CTCTTCCCCACGACCTCAGGGACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615826 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIPAL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NIPAL3 - NIPA like domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322854.1 | 136 | UTR 5 | NP_001309783.1 | |||
NM_001322855.1 | 136 | Intron | NP_001309784.1 | |||
NM_001322856.1 | 136 | Intron | NP_001309785.1 | |||
NM_001322857.1 | 136 | Intron | NP_001309786.1 | |||
NM_001322858.1 | 136 | Intron | NP_001309787.1 | |||
NM_001322859.1 | 136 | Intron | NP_001309788.1 | |||
NM_001322860.1 | 136 | Intron | NP_001309789.1 | |||
NM_001322861.1 | 136 | Intron | NP_001309790.1 | |||
NM_001322862.1 | 136 | Intron | NP_001309791.1 | |||
NM_001322863.1 | 136 | Intron | NP_001309792.1 | |||
NM_001322864.1 | 136 | Intron | NP_001309793.1 | |||
NM_001322865.1 | 136 | Intron | NP_001309794.1 | |||
NM_001322866.1 | 136 | Intron | NP_001309795.1 | |||
NM_001322868.1 | 136 | Intron | NP_001309797.1 | |||
NM_020448.4 | 136 | Intron | NP_065181.1 | |||
XM_011541808.2 | 136 | UTR 5 | XP_011540110.1 | |||
XM_017001868.1 | 136 | Intron | XP_016857357.1 | |||
XM_017001869.1 | 136 | Intron | XP_016857358.1 | |||
XM_017001870.1 | 136 | Intron | XP_016857359.1 |
STPG1 - sperm tail PG-rich repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199012.1 | 136 | Intron | NP_001185941.1 | |||
NM_001199013.1 | 136 | Intron | NP_001185942.1 | |||
NM_001199014.1 | 136 | Intron | NP_001185943.1 | |||
NM_178122.4 | 136 | Intron | NP_835223.1 | |||
XM_006711025.2 | 136 | Intron | XP_006711088.1 | |||
XM_011542403.2 | 136 | Intron | XP_011540705.1 | |||
XM_011542404.1 | 136 | Intron | XP_011540706.1 | |||
XM_017002768.1 | 136 | Intron | XP_016858257.1 | |||
XM_017002769.1 | 136 | Intron | XP_016858258.1 | |||
XM_017002770.1 | 136 | Intron | XP_016858259.1 |
Set Membership: |
HapMap |