Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCCACTTTCTGGATCGCCGAC[G/T]CTCTTGCCTTGGTCCGCTTGGGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOOK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOOK1 - hook microtubule-tethering protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015888.4 | Intron | NP_056972.1 | ||||
XM_006710676.1 | Intron | XP_006710739.1 | ||||
XM_011541562.2 | Intron | XP_011539864.1 | ||||
XM_011541563.1 | Intron | XP_011539865.1 | ||||
XM_017001424.1 | Intron | XP_016856913.1 |