Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602525 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDK2 - pyruvate dehydrogenase kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199898.1 | 4073 | Intron | NP_001186827.1 | |||
NM_001199899.1 | 4073 | Intron | NP_001186828.1 | |||
NM_001199900.1 | 4073 | Intron | NP_001186829.1 | |||
NM_002611.4 | 4073 | Intron | NP_002602.2 |
SAMD14 - sterile alpha motif domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257359.1 | 4073 | UTR 3 | NP_001244288.1 | |||
NM_174920.3 | 4073 | UTR 3 | NP_777580.1 | |||
XM_011524490.2 | 4073 | UTR 3 | XP_011522792.1 | |||
XM_017024322.1 | 4073 | UTR 3 | XP_016879811.1 | |||
XM_017024323.1 | 4073 | UTR 3 | XP_016879812.1 | |||
XM_017024324.1 | 4073 | UTR 3 | XP_016879813.1 |
Set Membership: |
HapMap |