Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATTTGATTTATTTCTTTTGGGT[A/G]GAATATAGGACTAGGGTAAGACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604918 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCLO PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCLO - piccolo presynaptic cytomatrix protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014510.2 | 17527 | Intron | NP_055325.2 | |||
NM_033026.5 | 17527 | UTR 3 | NP_149015.2 | |||
XM_017012006.1 | 17527 | UTR 3 | XP_016867495.1 | |||
XM_017012007.1 | 17527 | Intron | XP_016867496.1 |