Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTTCTCCCCAGTGAGGTTGTTG[A/G]GAAAAATTTAATCAGCTACAGATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300380 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTPS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
CTPS2 - CTP synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144002.1 | 2762 | UTR 3 | NP_001137474.1 | |||
NM_019857.4 | 2762 | UTR 3 | NP_062831.3 | |||
NM_175859.2 | 2762 | UTR 3 | NP_787055.1 | |||
XM_005274562.3 | 2762 | UTR 3 | XP_005274619.1 | |||
XM_006724503.3 | 2762 | UTR 3 | XP_006724566.1 | |||
XM_011545545.2 | 2762 | UTR 3 | XP_011543847.1 |
Set Membership: |
HapMap Validated |