Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACCTCTCTGCCCTATGATTTTC[C/T]GTAATTTTCGATCATCTATGCATTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611098 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STEAP4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
STEAP4 - STEAP4 metalloreductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205315.1 | 2669 | UTR 3 | NP_001192244.1 | |||
NM_001205316.1 | 2669 | UTR 3 | NP_001192245.1 | |||
NM_024636.3 | 2669 | UTR 3 | NP_078912.2 |