Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614144 MIM: 602521 MIM: 600596 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B9D1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
B9D1 - B9 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK7 - mitogen-activated protein kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002749.3 | 1131 | Intron | NP_002740.2 | |||
NM_139032.2 | 1131 | Intron | NP_620601.1 | |||
NM_139033.2 | 1131 | Intron | NP_620602.2 | |||
NM_139034.2 | 1131 | Intron | NP_620603.2 | |||
XM_006721557.3 | 1131 | Intron | XP_006721620.1 | |||
XM_006721558.3 | 1131 | Intron | XP_006721621.1 | |||
XM_006721559.3 | 1131 | Intron | XP_006721622.1 | |||
XM_011523957.2 | 1131 | Intron | XP_011522259.1 |
MFAP4 - microfibrillar associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198695.1 | 1131 | UTR 3 | NP_001185624.1 | |||
NM_002404.2 | 1131 | UTR 3 | NP_002395.1 |
Set Membership: |
HapMap |