Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602989 MIM: 606913 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLK2 - CDC like kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294338.1 | Intron | NP_001281267.1 | ||||
NM_001294339.1 | Intron | NP_001281268.1 | ||||
NM_003993.3 | Intron | NP_003984.2 | ||||
XM_005244878.1 | Intron | XP_005244935.1 | ||||
XM_011509143.1 | Intron | XP_011507445.1 | ||||
XM_011509144.1 | Intron | XP_011507446.1 | ||||
XM_011509147.2 | Intron | XP_011507449.1 | ||||
XM_017000241.1 | Intron | XP_016855730.1 | ||||
XM_017000242.1 | Intron | XP_016855731.1 | ||||
XM_017000243.1 | Intron | XP_016855732.1 | ||||
XM_017000244.1 | Intron | XP_016855733.1 |
FAM189B - family with sequence similarity 189 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAMP3 - secretory carrier membrane protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |