Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604692 MIM: 609475 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AKAP8 - A-kinase anchoring protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AKAP8L - A-kinase anchoring protein 8 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291478.1 | Intron | NP_001278407.1 | ||||
NM_014371.3 | Intron | NP_055186.3 | ||||
XM_005259854.3 | Intron | XP_005259911.2 | ||||
XM_011527916.2 | Intron | XP_011526218.1 |
Set Membership: |
HapMap Validated |