Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAAGTAATAATGTATGAACACC[A/G]TGTCCTTAACTTTTTATTCAGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607742 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KRT24 - keratin 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019016.2 | Intron | NP_061889.2 | ||||
XM_006721739.3 | Intron | XP_006721802.1 | ||||
XM_011524460.2 | Intron | XP_011522762.1 | ||||
XM_017024299.1 | Intron | XP_016879788.1 |
LOC105371775 - uncharacterized LOC105371775 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |