Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCATGGGAACTCAGTAGGTAG[G/A]TACAGCAAGGGGCAGTTTATTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153370 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITGAL - integrin subunit alpha L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114380.1 | Intron | NP_001107852.1 | ||||
NM_002209.2 | Intron | NP_002200.2 | ||||
XM_005255313.1 | Intron | XP_005255370.1 | ||||
XM_006721044.1 | Intron | XP_006721107.1 | ||||
XM_011545849.1 | Intron | XP_011544151.1 |
Set Membership: |
HapMap |