Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC14L6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SEC14L6 - SEC14 like lipid binding 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193336.2 | 1325 | Silent Mutation | GAA,GAG | E,E 349 | NP_001180265.2 | |
XM_011530357.2 | 1325 | Silent Mutation | GAA,GAG | E,E 366 | XP_011528659.1 | |
XM_011530358.2 | 1325 | Silent Mutation | GAA,GAG | E,E 334 | XP_011528660.1 | |
XM_011530360.2 | 1325 | Silent Mutation | GAA,GAG | E,E 303 | XP_011528662.1 | |
XM_017028931.1 | 1325 | Silent Mutation | GAA,GAG | E,E 384 | XP_016884420.1 | |
XM_017028932.1 | 1325 | Silent Mutation | GAA,GAG | E,E 295 | XP_016884421.1 | |
XM_017028933.1 | 1325 | Intron | XP_016884422.1 | |||
XM_017028934.1 | 1325 | Intron | XP_016884423.1 |