Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAAAAGGAAAAAGGAGAAAAAAC[C/T]CTAAATCAATGTTGCAAAGCAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CECR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CECR5 - cat eye syndrome chromosome region, candidate 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017829.5 | Intron | NP_060299.4 | ||||
NM_033070.2 | Intron | NP_149061.1 | ||||
XM_005261236.3 | Intron | XP_005261293.1 | ||||
XM_011546126.2 | Intron | XP_011544428.1 | ||||
XM_011546127.1 | Intron | XP_011544429.1 | ||||
XM_017028781.1 | Intron | XP_016884270.1 | ||||
XM_017028782.1 | Intron | XP_016884271.1 | ||||
XM_017028783.1 | Intron | XP_016884272.1 | ||||
XM_017028784.1 | Intron | XP_016884273.1 |
LOC100996342 - uncharacterized LOC100996342 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |