Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616747 MIM: 604759 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CHPT1 - choline phosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020244.2 | Intron | NP_064629.2 | ||||
XM_011538574.1 | Intron | XP_011536876.1 | ||||
XM_011538575.1 | Intron | XP_011536877.1 |
SYCP3 - synaptonemal complex protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177948.1 | Intron | NP_001171419.1 | ||||
NM_001177949.1 | Intron | NP_001171420.1 | ||||
NM_153694.4 | Intron | NP_710161.1 | ||||
XM_005268922.4 | Intron | XP_005268979.3 | ||||
XM_005268924.1 | Intron | XP_005268981.1 | ||||
XM_005268925.1 | Intron | XP_005268982.1 | ||||
XM_005268926.3 | Intron | XP_005268983.1 | ||||
XM_005268927.2 | Intron | XP_005268984.1 | ||||
XM_011538421.2 | Intron | XP_011536723.2 | ||||
XM_017019368.1 | Intron | XP_016874857.1 |
Set Membership: |
HapMap Validated |