Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 161565 MIM: 604028 MIM: 606473 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKTR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NKTR - natural killer cell triggering receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005385.3 | Intron | NP_005376.2 | ||||
XM_005265173.1 | Intron | XP_005265230.1 | ||||
XM_006713171.1 | Intron | XP_006713234.1 | ||||
XM_006713173.2 | Intron | XP_006713236.1 | ||||
XM_011533747.2 | Intron | XP_011532049.1 | ||||
XM_011533750.2 | Intron | XP_011532052.1 | ||||
XM_017006474.1 | Intron | XP_016861963.1 | ||||
XM_017006475.1 | Intron | XP_016861964.1 | ||||
XM_017006476.1 | Intron | XP_016861965.1 | ||||
XM_017006477.1 | Intron | XP_016861966.1 | ||||
XM_017006478.1 | Intron | XP_016861967.1 | ||||
XM_017006479.1 | Intron | XP_016861968.1 | ||||
XM_017006480.1 | Intron | XP_016861969.1 |
SEC22C - SEC22 homolog C, vesicle trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201572.1 | Intron | NP_001188501.1 | ||||
NM_001201584.1 | Intron | NP_001188513.1 | ||||
NM_004206.3 | Intron | NP_004197.1 | ||||
NM_032970.3 | Intron | NP_116752.1 | ||||
XM_005265533.1 | Intron | XP_005265590.1 | ||||
XM_005265534.1 | Intron | XP_005265591.1 | ||||
XM_005265535.1 | Intron | XP_005265592.1 | ||||
XM_011534212.1 | Intron | XP_011532514.1 | ||||
XM_017007438.1 | Intron | XP_016862927.1 | ||||
XM_017007439.1 | Intron | XP_016862928.1 |
SS18L2 - SS18 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |