Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCCTTCCTCTCTGACAGGGTTT[A/G]GATTCTGGGCTCTCAGCCTTAAAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615507 MIM: 608560 | |||||||||||||||||||||||
Literature Links: |
NISCH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NISCH - nischarin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAB1 - stabilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015136.2 | Intron | NP_055951.2 | ||||
XM_005264973.3 | Intron | XP_005265030.1 | ||||
XM_005264974.2 | Intron | XP_005265031.1 | ||||
XM_006713065.1 | Intron | XP_006713128.1 | ||||
XM_017005998.1 | Intron | XP_016861487.1 | ||||
XM_017005999.1 | Intron | XP_016861488.1 | ||||
XM_017006000.1 | Intron | XP_016861489.1 | ||||
XM_017006001.1 | Intron | XP_016861490.1 | ||||
XM_017006002.1 | Intron | XP_016861491.1 | ||||
XM_017006003.1 | Intron | XP_016861492.1 | ||||
XM_017006004.1 | Intron | XP_016861493.1 |