Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601041 MIM: 612399 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TLE2 - transducin like enhancer of split 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144761.1 | 2481 | UTR 3 | NP_001138233.1 | |||
NM_001144762.1 | 2481 | Silent Mutation | NP_001138234.1 | |||
NM_001300846.1 | 2481 | Silent Mutation | NP_001287775.1 | |||
NM_003260.4 | 2481 | Silent Mutation | NP_003251.2 | |||
XM_006722864.2 | 2481 | UTR 3 | XP_006722927.1 | |||
XM_011528230.1 | 2481 | Silent Mutation | XP_011526532.1 |
TLE6 - transducin like enhancer of split 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |