Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCACAGGTCAGCCCTGATAGCT[C/G]AAGATGTAGCAGCCCCAGTACCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603507 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506248 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100506248 - prothymosin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRP6 - LDL receptor related protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002336.2 | 6578 | UTR 3 | NP_002327.2 | |||
XM_006719078.3 | 6578 | UTR 3 | XP_006719141.1 | |||
XM_011520671.2 | 6578 | UTR 3 | XP_011518973.1 | |||
XM_017019304.1 | 6578 | UTR 3 | XP_016874793.1 |
MIR1244-4 - microRNA 1244-4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |