Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTAGTTAAATTTATTTCAGTGA[A/G]ATACTTTTGGAATTAATGTCCTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF38B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRPF38B - pre-mRNA processing factor 38B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018061.2 | 455 | Intron | NP_060531.2 | |||
XM_011541687.2 | 455 | Intron | XP_011539989.1 | |||
XM_017001596.1 | 455 | Intron | XP_016857085.1 | |||
XM_017001597.1 | 455 | Intron | XP_016857086.1 | |||
XM_017001598.1 | 455 | UTR 5 | XP_016857087.1 |