Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604042 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD35 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ANKRD35 - ankyrin repeat domain 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGA10 - integrin subunit alpha 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303040.1 | 3089 | UTR 3 | NP_001289969.1 | |||
NM_001303041.1 | 3089 | UTR 3 | NP_001289970.1 | |||
NM_003637.4 | 3089 | UTR 3 | NP_003628.2 | |||
XM_005277436.3 | 3089 | UTR 3 | XP_005277493.1 | |||
XM_011510083.2 | 3089 | UTR 3 | XP_011508385.1 | |||
XM_011510084.1 | 3089 | UTR 3 | XP_011508386.1 | |||
XM_017002622.1 | 3089 | UTR 3 | XP_016858111.1 | |||
XM_017002623.1 | 3089 | UTR 3 | XP_016858112.1 | |||
XM_017002624.1 | 3089 | UTR 3 | XP_016858113.1 | |||
XM_017002625.1 | 3089 | UTR 3 | XP_016858114.1 | |||
XM_017002626.1 | 3089 | UTR 3 | XP_016858115.1 | |||
XM_017002627.1 | 3089 | Intron | XP_016858116.1 | |||
XM_017002628.1 | 3089 | UTR 3 | XP_016858117.1 |
Set Membership: |
HapMap JSNP Validated |