Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 193002 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC18A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC18A1 - solute carrier family 18 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135691.2 | 2492 | UTR 3 | NP_001129163.1 | |||
NM_001142324.1 | 2492 | UTR 3 | NP_001135796.1 | |||
NM_001142325.1 | 2492 | UTR 3 | NP_001135797.1 | |||
NM_003053.3 | 2492 | UTR 3 | NP_003044.1 | |||
XM_011544625.1 | 2492 | UTR 3 | XP_011542927.1 | |||
XM_011544626.1 | 2492 | UTR 3 | XP_011542928.1 | |||
XM_011544627.1 | 2492 | UTR 3 | XP_011542929.1 |
Set Membership: |
HapMap Validated |