Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607301 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRPF3 - pre-mRNA processing factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004698.2 | Intron | NP_004689.1 | ||||
XM_011510129.1 | Intron | XP_011508431.1 | ||||
XM_011510130.2 | Intron | XP_011508432.1 | ||||
XM_011510131.2 | Intron | XP_011508433.1 | ||||
XM_011510132.2 | Intron | XP_011508434.1 | ||||
XM_017002790.1 | Intron | XP_016858279.1 | ||||
XM_017002791.1 | Intron | XP_016858280.1 |
Set Membership: |
HapMap |