Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR78 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDR78 - WD repeat domain 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024763.4 | Intron | NP_079039.4 | ||||
NM_207014.2 | Intron | NP_996897.2 | ||||
XM_005271212.3 | Intron | XP_005271269.1 | ||||
XM_011542161.2 | Intron | XP_011540463.1 | ||||
XM_011542162.2 | Intron | XP_011540464.1 | ||||
XM_017002353.1 | Intron | XP_016857842.1 | ||||
XM_017002354.1 | Intron | XP_016857843.1 | ||||
XM_017002355.1 | Intron | XP_016857844.1 | ||||
XM_017002356.1 | Intron | XP_016857845.1 | ||||
XM_017002357.1 | Intron | XP_016857846.1 |
Set Membership: |
HapMap |