Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615794 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FNDC3A - fibronectin type III domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079673.1 | Intron | NP_001073141.1 | ||||
NM_001278438.1 | Intron | NP_001265367.1 | ||||
NM_014923.4 | Intron | NP_055738.3 | ||||
XM_011534997.2 | Intron | XP_011533299.1 | ||||
XM_017020440.1 | Intron | XP_016875929.1 | ||||
XM_017020441.1 | Intron | XP_016875930.1 | ||||
XM_017020442.1 | Intron | XP_016875931.1 | ||||
XM_017020443.1 | Intron | XP_016875932.1 |
Set Membership: |
HapMap Validated |