Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTGCGTGGAGACGATGATCCTG[A/C]CAGCTCCCTTTTGGAAATCGTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602516 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RGS4 - regulator of G-protein signaling 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102445.2 | 289 | UTR 5 | NP_001095915.1 | |||
NM_001113380.1 | 289 | Intron | NP_001106851.1 | |||
NM_001113381.1 | 289 | Intron | NP_001106852.1 | |||
NM_005613.5 | 289 | Intron | NP_005604.1 |
Set Membership: |
HapMap |