Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGACCCTGCTGGGATTTGAACCT[A/G]CAGTTTCTATTAAGTTTGGTGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603536 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FUBP3 - far upstream element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003934.1 | Intron | NP_003925.1 | ||||
XM_005272232.1 | Intron | XP_005272289.1 | ||||
XM_006717313.2 | Intron | XP_006717376.1 | ||||
XM_011519172.2 | Intron | XP_011517474.1 | ||||
XM_011519173.2 | Intron | XP_011517475.1 | ||||
XM_011519174.2 | Intron | XP_011517476.1 | ||||
XM_017015269.1 | Intron | XP_016870758.1 | ||||
XM_017015270.1 | Intron | XP_016870759.1 | ||||
XM_017015271.1 | Intron | XP_016870760.1 | ||||
XM_017015272.1 | Intron | XP_016870761.1 | ||||
XM_017015273.1 | Intron | XP_016870762.1 |
LOC100272217 - uncharacterized LOC100272217 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |