Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTTACTGCTGCGGAGCGCTTTC[C/G]TTCCCGAGAATCAGTAAGCCCCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC171 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC171 - coiled-coil domain containing 171 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173550.2 | 303 | Intron | NP_775821.2 | |||
XM_005251397.2 | 303 | Intron | XP_005251454.1 | |||
XM_005251398.3 | 303 | Intron | XP_005251455.1 | |||
XM_005251399.4 | 303 | Intron | XP_005251456.1 | |||
XM_011517788.2 | 303 | UTR 5 | XP_011516090.1 | |||
XM_011517791.2 | 303 | Intron | XP_011516093.1 | |||
XM_011517796.2 | 303 | Intron | XP_011516098.1 | |||
XM_017014431.1 | 303 | Intron | XP_016869920.1 | |||
XM_017014432.1 | 303 | Intron | XP_016869921.1 | |||
XM_017014433.1 | 303 | Intron | XP_016869922.1 | |||
XM_017014434.1 | 303 | Intron | XP_016869923.1 | |||
XM_017014435.1 | 303 | Intron | XP_016869924.1 | |||
XM_017014436.1 | 303 | Intron | XP_016869925.1 | |||
XM_017014437.1 | 303 | Intron | XP_016869926.1 | |||
XM_017014438.1 | 303 | Intron | XP_016869927.1 | |||
XM_017014439.1 | 303 | Intron | XP_016869928.1 | |||
XM_017014440.1 | 303 | Intron | XP_016869929.1 | |||
XM_017014441.1 | 303 | Intron | XP_016869930.1 | |||
XM_017014442.1 | 303 | Intron | XP_016869931.1 | |||
XM_017014443.1 | 303 | Intron | XP_016869932.1 | |||
XM_017014444.1 | 303 | Intron | XP_016869933.1 |