Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601051 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR662 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MIR662 - microRNA 662 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSLN - mesothelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177355.2 | Intron | NP_001170826.1 | ||||
NM_005823.5 | Intron | NP_005814.2 | ||||
NM_013404.4 | Intron | NP_037536.2 | ||||
XM_005255034.4 | Intron | XP_005255091.1 | ||||
XM_011522348.2 | Intron | XP_011520650.1 | ||||
XM_017022853.1 | Intron | XP_016878342.1 | ||||
XM_017022854.1 | Intron | XP_016878343.1 | ||||
XM_017022855.1 | Intron | XP_016878344.1 | ||||
XM_017022856.1 | Intron | XP_016878345.1 | ||||
XM_017022857.1 | Intron | XP_016878346.1 | ||||
XM_017022858.1 | Intron | XP_016878347.1 |
Set Membership: |
HapMap Validated |