Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTTTGATAAATGAAATTTTG[A/G]TACCAAGATGGTGTTGCACTAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300204 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MID2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MID2 - midline 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012216.3 | Intron | NP_036348.2 | ||||
NM_052817.2 | Intron | NP_438112.2 | ||||
XM_005262062.4 | Intron | XP_005262119.1 | ||||
XM_017029239.1 | Intron | XP_016884728.1 |
Set Membership: |
HapMap |