Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTAAAAAGAAGCTTGTTAAAATT[A/G]ATTTGGCACCACGCACAGTGGCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605126 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHL5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
FHL5 - four and a half LIM domains 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170807.2 | Intron | NP_001164278.1 | ||||
NM_001322466.1 | Intron | NP_001309395.1 | ||||
NM_001322467.1 | Intron | NP_001309396.1 | ||||
NM_020482.5 | Intron | NP_065228.4 |
Set Membership: |
HapMap |