Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTGTAGAGGAAGTATTAATAAT[G/T]TTTGAAGCCAAGCAGATAATTTAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600497 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929935 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929935 - uncharacterized LOC101929935 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAA2 - protein kinase AMP-activated catalytic subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006252.3 | Intron | NP_006243.2 | ||||
XM_017001692.1 | Intron | XP_016857181.1 | ||||
XM_017001693.1 | Intron | XP_016857182.1 | ||||
XM_017001694.1 | Intron | XP_016857183.1 |
Set Membership: |
HapMap |