Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606610 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSFL1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NSFL1C - NSFL1 cofactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206736.1 | 3399 | UTR 3 | NP_001193665.1 | |||
NM_016143.4 | 3399 | UTR 3 | NP_057227.2 | |||
NM_018839.4 | 3399 | UTR 3 | NP_061327.2 | |||
XM_006723592.2 | 3399 | UTR 3 | XP_006723655.1 | |||
XM_006723593.2 | 3399 | UTR 3 | XP_006723656.1 | |||
XM_006723594.2 | 3399 | UTR 3 | XP_006723657.1 | |||
XM_011529300.1 | 3399 | UTR 3 | XP_011527602.1 | |||
XM_011529301.1 | 3399 | UTR 3 | XP_011527603.1 | |||
XM_011529302.2 | 3399 | UTR 3 | XP_011527604.1 | |||
XM_011529304.1 | 3399 | UTR 3 | XP_011527606.1 | |||
XM_011529305.2 | 3399 | Intron | XP_011527607.1 | |||
XM_017027956.1 | 3399 | UTR 3 | XP_016883445.1 | |||
XM_017027957.1 | 3399 | UTR 3 | XP_016883446.1 | |||
XM_017027958.1 | 3399 | UTR 3 | XP_016883447.1 |
Set Membership: |
HapMap |