Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601505 MIM: 614275 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF146 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF146 - zinc finger protein 146 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099638.1 | Intron | NP_001093108.1 | ||||
NM_001099639.1 | Intron | NP_001093109.1 | ||||
NM_007145.2 | Intron | NP_009076.2 | ||||
XM_005259214.2 | Intron | XP_005259271.1 | ||||
XM_017027244.1 | Intron | XP_016882733.1 | ||||
XM_017027245.1 | Intron | XP_016882734.1 | ||||
XM_017027246.1 | Intron | XP_016882735.1 | ||||
XM_017027247.1 | Intron | XP_016882736.1 |
ZNF565 - zinc finger protein 565 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |